Learn More
TISAB IV solution, For determination of fluoride, MilliporeSigma™ Supelco™
Total ionic strength adjustment buffers (TISAB), is generally used for the microdetermination of fluorides.
Supplier:  MilliporeSigma Supelco 34574500ML
 View more versions of this product
Catalog No. 11-100-2586
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
64-19-7
Determination of Fluoride
500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories