Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TISAB, Low Level, Reagents

Catalog No. 01333432
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-432 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-432 Supplier Reagents Holdings Llc Supplier No. CT1302004A
Add to Cart
Add to Cart

TISAB, Low Level, Reagent, use for Water analysis (Fluoride). Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7 , 7647-14-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Total Ionic Strength Adjustment Buffer
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
For Use With (Equipment) Fluoride Meter
pH 5 to 5.5
Physical Form Liquid
Quantity 4 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.0483
Sterility Non-sterile
Grade Reagent
Chemical Name or Material TISAB, Low Level
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.