Learn More
Tisch Scientific Vial Rack for 27.5 mL Vials, White Polypropylene, 10.83" x 6.85" x 1.22", 40 Holes, Hole DIameter: 29 mm, Hole Depth: 24 mm, Non-Autoclavable, 1 Pack
SDP
Catalog No. 502251503
Encompass_Preferred
Tisch 2mL vial racks are designed for use in laboratories, medical settings, research and more. Vial racks are made from industry-standard polypropylene plastic and hold up to 50 vials per rack.
This item is not returnable. View return policy
Catalog No. 50-225-1503 Supplier Tisch Scientific Supplier No. CV20561
Tisch 2mL vial racks are designed for use in laboratories, medical settings, research and more. Vial racks are made from industry-standard polypropylene plastic and hold up to 50 vials per rack.