Learn More
Tisch Scientific CELLULOSE ACETATE SYRINGE
SDP
Supplier:  Tisch Scientific SF14488
Encompass
Tisch cellulose acetate syringe filters are a hydrophilic filtration option perfect for use in aqueous protein solutions and ground water filtration applications.
Catalog No. 50-225-1452