Learn More
Tisch Scientific GLASS FIBER SYRINGE FILTER
SDP
Supplier:  Tisch Scientific SF14699
Encompass
Tisch glass fiber syringe filters are ideal for the filtration of extremely viscous samples which could clog other standard syringe filters. These hydrophilic syringe filters are known for their excellent compatibility with organic solvents, strong acids and bases
Catalog No. 50-225-1453