Learn More
Tisch Scientific MCE Membrane Filters
SDP
Supplier:  Tisch Scientific SF14709
Encompass
Tisch MCE membrane filters are developed from a mixture of cellulose acetate and cellulose nitrate and are one of the most widely used filters in both air monitoring and analytical applications. These filters are gridded for microbial cell counting and particle detection.
Catalog No. 50-225-1439