Learn More
Tisch Scientific NYLON MEMBRANE FILTERS
SDP
Supplier:  Tisch Scientific SF14530
Encompass
Tisch nylon membrane filters are naturally hydrophilic, have high non-specific binding properties, and an extractable level of <0.0015 mg/cm^2, they are ideal for HPLC solvent and sample preparation.
Catalog No. 50-225-1434