Learn More
Tisch Scientific NYLON SYRINGE FILTERS
SDP
Supplier:  Tisch Scientific SF14478
Encompass
Tisch nylon syringe filters provide a universal application for analytical procedures. Naturally hydrophilic, high protein binding and high dirt loading capability, and good chemical resistance make them ideal for aqueous (non-acidic) or organic sample preparations, HPLC, GC or dissolution sample analysis.
Catalog No. 50-225-1456