Learn More
Tisch Scientific PTFE SYRINGE FILTERS
SDP
Supplier:  Tisch Scientific SF14468
Encompass
Tisch PTFE Syringe filters are very versatile, proving to be effective for applications such as alkali filtration, strong acid solvent filtration, and organic solvent with strong chemical causticity filtration.
Catalog No. 50-225-1466