Learn More
Tisch Scientific SCIENTIFIC POLYTETRAFLUO
SDP
Catalog No. 502251423
Encompass
Tisch Scientific polytetrafluoroethylene (PFTE) membrane filters from Tisch are made to both sterilize gas and clarify liquids in lab environments. Their PTFE membranes are hydrophobic and resistant to aggressive media and corrosive solvents, making them ideal for a variety of filtration applications.
This item is not returnable. View return policy
Catalog No. 50-225-1423 Supplier Tisch Scientific Supplier No. SF16015
Tisch Scientific polytetrafluoroethylene (PFTE) membrane filters from Tisch are made to both sterilize gas and clarify liquids in lab environments. Their PTFE membranes are hydrophobic and resistant to aggressive media and corrosive solvents, making them ideal for a variety of filtration applications.