Learn More
Tisch Scientific Polytetrafluoroethylene (PFTE) Membrane Filters
SDP
Supplier:  Tisch Scientific SF18116
Encompass
Tisch Scientific polytetrafluoroethylene (PFTE) membrane filters from Tisch are made to both sterilize gas and clarify liquids in lab environments. Their PTFE membranes are hydrophobic and resistant to aggressive media and corrosive solvents, making them ideal for a variety of filtration applications.
Catalog No. 50-225-1494