Learn More
Sklar Instruments Tischler-Kevorkian Biopsy Punch, Sklar, 12 in., with Locking Tip, Straight, Rectangular Bite with Upper and Lower Teeth, Stainless Steel. EA/1.
SDP
Catalog No. 501436834 Shop All Sklar Instruments Products
Encompass_Preferred
Tischler-Kevorkian Biopsy Punch, Sklar, 12 in., with Locking Tip, Straight, Rectangular Bite with Upper and Lower Teeth, Stainless Steel. Sklar's Tischler-Kevorkian Biopsy Punch is an OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy. This product is straight with an 8 inch shaft and 3mm x 9.5mm rectangular bite. It features a single lower tooth and a single upper tooth used to grasp difficult to reach tissue. It also has a locking mechanism above the horn on the handle so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal. This reduces the risk of losing the tissue sample as the instrument is withdrawn. This product has a pistol style handle with one large ring for multiple fingers.
This item is not returnable. View return policy
Catalog No. 50-143-6834 Supplier Sklar Instruments Supplier No. 906887
Tischler-Kevorkian Biopsy Punch, Sklar, 12 in., with Locking Tip, Straight, Rectangular Bite with Upper and Lower Teeth, Stainless Steel. Sklar's Tischler-Kevorkian Biopsy Punch is an OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy. This product is straight with an 8 inch shaft and 3mm x 9.5mm rectangular bite. It features a single lower tooth and a single upper tooth used to grasp difficult to reach tissue. It also has a locking mechanism above the horn on the handle so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal. This reduces the risk of losing the tissue sample as the instrument is withdrawn. This product has a pistol style handle with one large ring for multiple fingers.