Learn More
Medchemexpress LLC Tisolagiline monomethylsulfate | 1894207-45-4 | 99.8% | 10 MG
SDP
Catalog No. 5000339634
Encompass_Preferred
Tisolagiline monomethylsulfate (KDS2010) is a selective, reversible, and orally active inhibitor for MAO-B with an IC50 of 8 nM. It exhibits neuroprotective and anti-neuroinflammatory activities. It reduces glial fibrillary acidic protein (GFAP) expression and glycosaminoglycan (CSPGs) deposition, thereby inhibiting astrocyte activation and scar formation.
  • Ameliorates MPTP-induced Parkinson's disease in mouse models, alleviating motor dysfunction.
  • Reverses reduction in TH-positive neurons and reduces GFAP and iNOS expression.
  • Inhibits astrocyte scar formation.
  • Increases doxorubicin penetration into glioblastoma domains and glioblastoma tumor volume in mouse models.
Catalog No. 50-003-39634 Supplier Medchemexpress LLC Supplier No. HY127109A10MG
Tisolagiline monomethylsulfate (KDS2010) is a selective, reversible, and orally active inhibitor for MAO-B with an IC50 of 8 nM. It exhibits neuroprotective and anti-neuroinflammatory activities. It reduces glial fibrillary acidic protein (GFAP) expression and glycosaminoglycan (CSPGs) deposition, thereby inhibiting astrocyte activation and scar formation.
  • Ameliorates MPTP-induced Parkinson's disease in mouse models, alleviating motor dysfunction.
  • Reverses reduction in TH-positive neurons and reduces GFAP and iNOS expression.
  • Inhibits astrocyte scar formation.
  • Increases doxorubicin penetration into glioblastoma domains and glioblastoma tumor volume in mouse models.