Learn More
Medchemexpress LLC Tisolagiline monomethylsulfate | 1894207-45-4 | 99.8% | 418.43 | C18H21F3N2O4S | 5 MG
SDP
Catalog No. 5000264089
Encompass_Preferred
Tisolagiline monomethylsulfate is a selective, reversible, orally active monoamine oxidase B (MAO-B) inhibitor used in preclinical research for neuroprotection and anti-neuroinflammatory studies. It is supplied as a white to off-white solid with formula C18H21F3N2O4S and molecular weight 418.43.
  • Selective, reversible MAO-B inhibitor with low-nanomolar potency.
  • Exhibits neuroprotective and anti-neuroinflammatory activity.
  • White to off-white solid suitable for analytical and in vivo studies.
  • High overall and enantiomeric purity (reported ~99.8% and ee 100%).
  • Available in mg-scale research quantities for preclinical work.
  • Storage recommendations: powder -20°C for long term, 4°C for short term; in solvent, -80°C.
Catalog No. 50-002-64089 Supplier Medchemexpress LLC Supplier No. HY127109A5MG
Tisolagiline monomethylsulfate is a selective, reversible, orally active monoamine oxidase B (MAO-B) inhibitor used in preclinical research for neuroprotection and anti-neuroinflammatory studies. It is supplied as a white to off-white solid with formula C18H21F3N2O4S and molecular weight 418.43.
  • Selective, reversible MAO-B inhibitor with low-nanomolar potency.
  • Exhibits neuroprotective and anti-neuroinflammatory activity.
  • White to off-white solid suitable for analytical and in vivo studies.
  • High overall and enantiomeric purity (reported ~99.8% and ee 100%).
  • Available in mg-scale research quantities for preclinical work.
  • Storage recommendations: powder -20°C for long term, 4°C for short term; in solvent, -80°C.