Learn More
Medchemexpress LLC Tisolagiline monomethylsulfate | 1894207-45-4 | 99.8% | 50 MG
SDP
Catalog No. 5000339635
Encompass_Preferred
Tisolagiline methylsulfate (KDS2010) is a selective, reversible, and orally active inhibitor for MAO-B with an IC50 of 8 nM. It exhibits neuroprotective and anti-neuroinflammatory activities.
  • Selective, reversible, and orally active MAO-B inhibitor
  • Exhibits neuroprotective and anti-neuroinflammatory activities
  • Reduces expression of glial fibrillary acidic protein (GFAP)
  • Ameliorates MPTP-induced Parkinson's disease in mouse models
  • Inhibits astrocyte scar formation
Catalog No. 50-003-39635 Supplier Medchemexpress LLC Supplier No. HY127109A50MG
Tisolagiline methylsulfate (KDS2010) is a selective, reversible, and orally active inhibitor for MAO-B with an IC50 of 8 nM. It exhibits neuroprotective and anti-neuroinflammatory activities.
  • Selective, reversible, and orally active MAO-B inhibitor
  • Exhibits neuroprotective and anti-neuroinflammatory activities
  • Reduces expression of glial fibrillary acidic protein (GFAP)
  • Ameliorates MPTP-induced Parkinson's disease in mouse models
  • Inhibits astrocyte scar formation