Learn More
Selleck Chemical LLC Tisotumab vedotin-D4051-1MG
SDP
Catalog No. 503136654
Encompass_Preferred
Tisotumab vedotin (TF-011-MMAE HuMax-TF-ADC) is an antibody-drug conjugate (ADC) composed of a fully human monoclonal antibody to tissue factor III (thromboplastin) also known as CD142 conjugated to Monomethyl Auristatin E (MMAE) via a cleavable maleimidocaproyl-valyl-citrullinylp-aminobenzyloxycarbonyl (mc-val-cit-PABC) type linker Tisotumab vedotin has the potential for use in research of solid tumors mainly in cervical cancer
Catalog No. 50-313-6654 Supplier Selleck Chemical LLC Supplier No. D40511MG
Tisotumab vedotin (TF-011-MMAE HuMax-TF-ADC) is an antibody-drug conjugate (ADC) composed of a fully human monoclonal antibody to tissue factor III (thromboplastin) also known as CD142 conjugated to Monomethyl Auristatin E (MMAE) via a cleavable maleimidocaproyl-valyl-citrullinylp-aminobenzyloxycarbonyl (mc-val-cit-PABC) type linker Tisotumab vedotin has the potential for use in research of solid tumors mainly in cervical cancer