Learn More
Andwin Scientific Tissue-Tek® O.C.T. Compound 4OZ
SDP
Catalog No. NC1029572
Encompass_Preferred
Tissue-Tek optimum cutting temperature O.C.T. Compound Provides a convenient specimen matrix for cryostat sectioning at temperatures of -10[degree]C and below. Made of water-soluble glycols and resins. It is packaged and sold individually in a 118mL (4oz.) bottle.
This item is not returnable. View return policy
Catalog No. NC1029572 Supplier Andwin Scientific Supplier No. 25608930
Tissue-Tek optimum cutting temperature O.C.T. Compound Provides a convenient specimen matrix for cryostat sectioning at temperatures of -10[degree]C and below. Made of water-soluble glycols and resins. It is packaged and sold individually in a 118mL (4oz.) bottle.