Learn More
Electron Microscopy Sciences Tissue Adhesive 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 72590
Encompass_Preferred
Permabond 910 contains cyanoacrylate ester. Immediate bonding of biological specimens. It is not water soluble. Used for mounting specimens for Vibratome sectioning.
Catalog No. 50-948-963