Learn More
Med Vet International Tissue Adhesive, 5 Seconds, Vet Use, Bottle, 3 G, Each
Catalog No. 501180910
Encompass_Preferred
Unscrew cap from 3mL bottle. Remove an applicator and fix to the exposed spout of the bottle. Holding the applicator above the incision line, apply slight pressure, thus releasing the adhesive in a thin line, while pinching the opposed line to maintain closure. Do not over apply as a thin application will be sufficient. Allow the topical skin adhesive to fully polymerize, which will take approximately 5 seconds. Compare to ZipStitch, Dermabond , Vetbond ™
Catalog No. 50-118-0910 Supplier Med Vet International Supplier No. TA5
 Restricted Item
Add to Cart
Add to Cart
Unscrew cap from 3mL bottle. Remove an applicator and fix to the exposed spout of the bottle. Holding the applicator above the incision line, apply slight pressure, thus releasing the adhesive in a thin line, while pinching the opposed line to maintain closure. Do not over apply as a thin application will be sufficient. Allow the topical skin adhesive to fully polymerize, which will take approximately 5 seconds. Compare to ZipStitch, Dermabond , Vetbond ™