Learn More
Electron Microscopy Sciences Tissue basket with Lid, 15 mm diameter, 10 mm height
SDP
Catalog No. 501926616
Encompass_Preferred
EMS has introduced and is now available a variety of Stainless steel baskets with lids and multiple compartments for critical point Drying. These baskets are either 15 mm in diameter or 23 mm.
Diameter: 15 mm
Height: 10 mm
Catalog No. 50-192-6616 Supplier Electron Microscopy Sciences Supplier No. 7019426
EMS has introduced and is now available a variety of Stainless steel baskets with lids and multiple compartments for critical point Drying. These baskets are either 15 mm in diameter or 23 mm.
Diameter: 15 mm
Height: 10 mm