Learn More
Electron Microscopy Sciences Tissue Basket with Lid, 25 mm diameter, 23 mm height
SDP
Catalog No. 501926617
Encompass_Preferred
EMS has introduced and is now available a variety of Stainless steel baskets with lids and multiple compartments for critical point Drying. These baskets are either 15 mm in diameter or 23 mm.
Diameter: 25 mm
Height: 23 mm
Catalog No. 50-192-6617 Supplier Electron Microscopy Sciences Supplier No. 7019427
EMS has introduced and is now available a variety of Stainless steel baskets with lids and multiple compartments for critical point Drying. These baskets are either 15 mm in diameter or 23 mm.
Diameter: 25 mm
Height: 23 mm