Learn More
Electron Microscopy Sciences QuantomiX WetSEM QX -302 Tissue Capsules
SDP
Catalog No. 5031337
Encompass_Preferred
The QX-302 Capsules are used for imaging various thick, non-adherent samples, such as tissue biopsies, plants and material specimens, in their natural wet state. The capsule is suitable for variable sample sizes, with a maximum diameter of 3 mm and thickness of up to 1 mm. No coating or embedding of the sample are required, enabling electron microscopy imaging with easy sample preparation comparable to light microscopy.
Catalog No. 50-313-37 Supplier Electron Microscopy Sciences Supplier No. QX3026
The QX-302 Capsules are used for imaging various thick, non-adherent samples, such as tissue biopsies, plants and material specimens, in their natural wet state. The capsule is suitable for variable sample sizes, with a maximum diameter of 3 mm and thickness of up to 1 mm. No coating or embedding of the sample are required, enabling electron microscopy imaging with easy sample preparation comparable to light microscopy.