Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCI America™ Tissue-Clearing Reagent iTOMEI-D
SDP

Catalog No. T394025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Capacity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
T394025ML 25 mL
T39405ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T394025ML Supplier TCI America™ Supplier No. T394025ML
Add to Cart
Add to Cart

Improved TOMEI (iTOMEI) is a method that specializes the TOMEI clearing process for fluorescent protein observation.

  • Enables transparency by simple osmosis in just a few days
  • Keeps the fluorescence of a fluorescent protein such as GFP or tdTomato co-expressed as a reporter gene
  • Suppresses auto-fluorescence
  • Applicable to wide variety of plant species including Oryza sativa, Arabidopsis thaliana, Marchantia polymorpha

Specifications

Capacity 25 mL
For Use With (Equipment) Confocal Laser-scanning Microscopy (CLSM) Device
Product Line TOMEI
For Use With (Application) Plant Cytology and Histological research
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.