Learn More
Electron Microscopy Sciences Swingsette™ Tissue Cassettes in QuickLoad Stacks, Gray
SDP
Catalog No. 501930960
Encompass_Preferred
These cassettes will load in Leica and Sakura cassette labeling printers in one simple operation of loading the cassette sleeve in the magazine. In convenient stacks of 40 cassettes that save time and money
The specially designed cassettes differ by the hinge that holds the base and cover together. This hinge allows the Swingsette to be opened and closed as often as necessary.
Anterior printing area is at a 45 degree angle, offering an unobstructed view of the writing surface.
Color: Gray
Type: Slotted
This item is not returnable. View return policy
Catalog No. 50-193-0960 Supplier Electron Microscopy Sciences Supplier No. 7007981
These cassettes will load in Leica and Sakura cassette labeling printers in one simple operation of loading the cassette sleeve in the magazine. In convenient stacks of 40 cassettes that save time and money
The specially designed cassettes differ by the hinge that holds the base and cover together. This hinge allows the Swingsette to be opened and closed as often as necessary.
Anterior printing area is at a 45 degree angle, offering an unobstructed view of the writing surface.
Color: Gray
Type: Slotted