Learn More
Electron Microscopy Sciences Unisette™ Tissue Cassettes in QuickLoad™ Stacks, Peach
SDP
Catalog No. 501930785
Encompass_Preferred
Molded from a special high density acetal, these tissue cassettes for Leica and Sakura printers keep specimens safely submerged in liquid and are totally resistant to the chemical action of solvents used in histology laboratories. The efficient flow-through slots maximize fluid exchange and ensure proper drainage.
The one-piece snap-latch and hinge-lock design prevents early separation of base and lid and allows one-hand operation. Lids are attached in an open position for easy filling, but can be opened or closed as often as necessary and will always relock securely. The tab on the left front side makes opening easy. Lids are easily removed by pulling sideways. The anterior writing area is at a 35 degree angle.
Color: Peach
This item is not returnable. View return policy
Catalog No. 50-193-0785 Supplier Electron Microscopy Sciences Supplier No. 7007941
Molded from a special high density acetal, these tissue cassettes for Leica and Sakura printers keep specimens safely submerged in liquid and are totally resistant to the chemical action of solvents used in histology laboratories. The efficient flow-through slots maximize fluid exchange and ensure proper drainage.
The one-piece snap-latch and hinge-lock design prevents early separation of base and lid and allows one-hand operation. Lids are attached in an open position for easy filling, but can be opened or closed as often as necessary and will always relock securely. The tab on the left front side makes opening easy. Lids are easily removed by pulling sideways. The anterior writing area is at a 35 degree angle.
Color: Peach