Learn More
Electron Microscopy Sciences Histosette Tissue Cassettes in QuickLoad™ Stacks, White
SDP
Catalog No. 501930767
Encompass_Preferred
Molded from a special high density acetal,the acetal caseettes keep specimens safely submerged and are resistant to the chemical action of most solvents used in histology laboratories. The efficient flow-through slots maximize fluid exchange and ensure proper reagent drainage.
The one-piece disposable plastic cover is pre-installed on each cassette and eliminates the need for reusable steel lids. The cover can be opened and closed as often as necessary and will always relock, reducing the possibility of specimen loss. The anterior printing area is at a 45° angle and offers an unobstructed view of the writing surface. For Leica and Sakura printers.
Color: White
This item is not returnable. View return policy
Catalog No. 50-193-0767 Supplier Electron Microscopy Sciences Supplier No. 7007912
Molded from a special high density acetal,the acetal caseettes keep specimens safely submerged and are resistant to the chemical action of most solvents used in histology laboratories. The efficient flow-through slots maximize fluid exchange and ensure proper reagent drainage.
The one-piece disposable plastic cover is pre-installed on each cassette and eliminates the need for reusable steel lids. The cover can be opened and closed as often as necessary and will always relock, reducing the possibility of specimen loss. The anterior printing area is at a 45° angle and offers an unobstructed view of the writing surface. For Leica and Sakura printers.
Color: White