Learn More
Chemglass Life Sciences Flask, Tissue Culture 75cm2, 250mL, Treated, Vent Cap, Sterile
SDP
Catalog No. 501897586
Encompass
• Angled neck for easy access• High clarity, 100% virgin polystyrene• Frosted writing and clear graduations• Notched bottom for slip-free stacking• Sterilized by E Beam• Vented caps (with breathable membrane)• Clear lot number for batch traceability• Non-pyrogenic, DNase- and RNase-freeCell culture flasks are available in cell growth areas ranging from 25cm2 to 225cm2. Tissue culture treated with a vent cap. Vent caps with 0.22 μm hydrophobic filters for gas exchange.
Catalog No. 50-189-7586 Supplier Chemglass Life Sciences Supplier No. CGE2406075
• Angled neck for easy access• High clarity, 100% virgin polystyrene• Frosted writing and clear graduations• Notched bottom for slip-free stacking• Sterilized by E Beam• Vented caps (with breathable membrane)• Clear lot number for batch traceability• Non-pyrogenic, DNase- and RNase-freeCell culture flasks are available in cell growth areas ranging from 25cm2 to 225cm2. Tissue culture treated with a vent cap. Vent caps with 0.22 μm hydrophobic filters for gas exchange.