Learn More
Research Products International Corp Tissue Culture Flask, 50ml, 200/cs
SDP
Catalog No. 501977466
Encompass
Manufactured from ultra clear polystyrene for easy viewing of contents. Proprietary surface treating ensures optimum cell attachment and growth. Additional feature include graduated markings on both sides of flask, angled neck design and vented caps with 0.22um filter. Supplied sterile, non-pyrogenic, RNase, DNase free.
Catalog No. 50-197-7466 Supplier Research Products International Corp Supplier No. 162431
Manufactured from ultra clear polystyrene for easy viewing of contents. Proprietary surface treating ensures optimum cell attachment and growth. Additional feature include graduated markings on both sides of flask, angled neck design and vented caps with 0.22um filter. Supplied sterile, non-pyrogenic, RNase, DNase free.