Learn More
Research Products International Corp Tissue Culture Flask Rack, Holds 12x75ml Flasks
SDP
Catalog No. 501978025
Encompass
Racks hold culture flasks open and ready for filling, aspirating, collecting, seeding cells, adding solutions and more. Culture flasks stand in a vertical position within the rack and are easily transported. Grips on each end of rack facilitate easy handling.
Catalog No. 50-197-8025 Supplier Research Products International Corp Supplier No. 246250
Racks hold culture flasks open and ready for filling, aspirating, collecting, seeding cells, adding solutions and more. Culture flasks stand in a vertical position within the rack and are easily transported. Grips on each end of rack facilitate easy handling.