Learn More
LAMDA BIOTECH INC TISSUE-DIRECT™ PCR Genotyping Kit
Supplier:  LAMDA BIOTECH INC D300100
Encompass_Preferred
Tissue-DirectTM PCR Genotyping Kit contains all the reagents needed for quick preparation of genomic DNA and genotyping by direct PCR amplification. Tissues used for this kit can be mouse tail and any other animal tissues. This kit can be used for other biological samples, such as plant, blood or bacteria with minor modifications. The PCR product can be directly loaded onto agarose gel without addition of loading dye for visualization of the experimental results.
Catalog No. 50-204-5785