Learn More
STEMCELL Technologies Tissue Dissociation Flask, Size: 1 Unit
SDP
Catalog No. 501974751
Encompass
Borosilicate glass flat-bottom 250 mL flask with break-resistant lip. Designed for the enzymatic digestion of tissue fragments and homogenates into cell suspensions.
Catalog No. 50-197-4751 Supplier STEMCELL Technologies Supplier No. 27300
Borosilicate glass flat-bottom 250 mL flask with break-resistant lip. Designed for the enzymatic digestion of tissue fragments and homogenates into cell suspensions.