Learn More
Alkali Scientific Tissue Embedding Cassette with Attached (Detachable) Lid 35° Writing Area, Dispenser Box, 500/Pk, Orange
SDP
Catalog No. 5000662057
Encompass_Preferred
These 35° angled Mega Type tissue embedding cassettes are specifically designed to accommodate medium to large tissue samples. The cassettes feature a one piece design with an attached lid that allows for repeated opening and closing without detachment, ensuring long term durability and convenience during sample processing. A secure locking mechanism keeps the lid tightly closed, maintaining sample integrity and safety. The large, textured writing surface and dual side panels provide ample space for easy labeling, while a front thumb tab ensures effortless opening. These cassettes are compatible with most manual cassette printers and are ideal for laboratories requiring reliable sample tracking and handling. Designed for medium to large tissue samples 35° angled design compatible with most cassette printers One piece construction with attached lid for repeated opening and closing Secure lid locking mechanism to protect samples Large textured writing surface and side panels for labelin
Catalog No. 50-006-62057 Supplier Alkali Scientific Supplier No. ES3013O
These 35° angled Mega Type tissue embedding cassettes are specifically designed to accommodate medium to large tissue samples. The cassettes feature a one piece design with an attached lid that allows for repeated opening and closing without detachment, ensuring long term durability and convenience during sample processing. A secure locking mechanism keeps the lid tightly closed, maintaining sample integrity and safety. The large, textured writing surface and dual side panels provide ample space for easy labeling, while a front thumb tab ensures effortless opening. These cassettes are compatible with most manual cassette printers and are ideal for laboratories requiring reliable sample tracking and handling. Designed for medium to large tissue samples 35° angled design compatible with most cassette printers One piece construction with attached lid for repeated opening and closing Secure lid locking mechanism to protect samples Large textured writing surface and side panels for labelin