Learn More
Alkali Scientific Tissue Embedding Cassette, Pink, 500 pk
SDP
Catalog No. 502389382
Encompass_Preferred
Tissue embedding cassettes Mega type for medium-big dimensions samples. Mega type tissue embedding cassettes are provided with clip on closure lid that can easily be removed. Product Dimensions (l x w x h): 1.1” x 1.6” x 0.54”.
Catalog No. 50-238-9382 Supplier Alkali Scientific Supplier No. ES3013
Tissue embedding cassettes Mega type for medium-big dimensions samples. Mega type tissue embedding cassettes are provided with clip on closure lid that can easily be removed. Product Dimensions (l x w x h): 1.1” x 1.6” x 0.54”.