Learn More
Meridian Life Science Tissue Extract-PCR Buffers, 1ML
SDP
Catalog No. 5000856387
Encompass_Preferred
Tissue Extract-PCR Buffer offers a convenient, fast and efficient method for the extraction of DNA from a variety of mammalian tissues, particularly from rodent tail or ear samples. The DNA extractions are performed in a single tube, without the need for multiple washing steps, greatly reducing the risk of sample loss and contamination, and can be used directly in a PCR reaction. Use with high quality Taq HS DNA Polymerase (MDX008) or inhibitor tolerant qPCR mixes for improving yield of even the most challenging targets
Catalog No. 50-008-56387 Supplier Meridian Life Science Supplier No. MDX0041
Tissue Extract-PCR Buffer offers a convenient, fast and efficient method for the extraction of DNA from a variety of mammalian tissues, particularly from rodent tail or ear samples. The DNA extractions are performed in a single tube, without the need for multiple washing steps, greatly reducing the risk of sample loss and contamination, and can be used directly in a PCR reaction. Use with high quality Taq HS DNA Polymerase (MDX008) or inhibitor tolerant qPCR mixes for improving yield of even the most challenging targets