Learn More
CPC Scientific H-Val-Tyr-Thr-Val-Gln-Ile-Cys-Thr-Lys-Ser-Gly-Asp-Trp-Lys-Ser-Lys-Cys-Phe-Tyr-Thr-Thr-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110557
Encompass
Tissue factor (TF) is a transmembrane protein, enables cells to initiate the blood coagulation cascades in vivo, functions as the high-affinity receptor for the coagulation factor VII. This peptide specially inhibit theTF dependent clotting cascade.ONE-LETTER SEQUENCE: VYTVQICTKSGDWKSKCFYTT(Cys&Cys bridge)MOLECULAR FORMULA: C111H166N26O33S2MOLECULAR WEIGHT:2456.83STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [313642-01-2], RESEARCH AREA: CardiovascularREFERENCES: L.R.Paborsky et al., Biochemistry, 34, 15328 (1995)
Catalog No. 50-211-0557 Supplier CPC Scientific Supplier No. TF001B
Tissue factor (TF) is a transmembrane protein, enables cells to initiate the blood coagulation cascades in vivo, functions as the high-affinity receptor for the coagulation factor VII. This peptide specially inhibit theTF dependent clotting cascade.ONE-LETTER SEQUENCE: VYTVQICTKSGDWKSKCFYTT(Cys&Cys bridge)MOLECULAR FORMULA: C111H166N26O33S2MOLECULAR WEIGHT:2456.83STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [313642-01-2], RESEARCH AREA: CardiovascularREFERENCES: L.R.Paborsky et al., Biochemistry, 34, 15328 (1995)