Learn More
Electron Microscopy Sciences Tissue File Case, Embedment Block Capacity: 100 to 1000 per tissue file, 6/pk
SDP
Catalog No. 5029327
Encompass_Preferred
A histological embedment block cataloging and storage system that combines embedment block protection, simplified cataloging, reduction of filing error and facilitation of retrieval. All of this while conserving space and being very affordable.
By removing alternating partitions, each compartment can be doubled or quadrupled in size. For other storage needs, the partitions may also be removed.
For cataloging, each compartment is numerically indexed on a permanent chart inside the lid which also provides space for detailed information. Tissue files are made from solid fiberboard with a protective, scuff-resistant exterior green covering. Its hinged lid lifts open and with a cloth stay support the lid can be kept in the open position.
Capacity is 100 to 1000 per tissue file. There are 100 of 1.5"x 1 x2.25" (38x25x57 mm) compartments arranged 10 across x 10 deep.
Catalog No. 50-293-27 Supplier Electron Microscopy Sciences Supplier No. 6328606
A histological embedment block cataloging and storage system that combines embedment block protection, simplified cataloging, reduction of filing error and facilitation of retrieval. All of this while conserving space and being very affordable.
By removing alternating partitions, each compartment can be doubled or quadrupled in size. For other storage needs, the partitions may also be removed.
For cataloging, each compartment is numerically indexed on a permanent chart inside the lid which also provides space for detailed information. Tissue files are made from solid fiberboard with a protective, scuff-resistant exterior green covering. Its hinged lid lifts open and with a cloth stay support the lid can be kept in the open position.
Capacity is 100 to 1000 per tissue file. There are 100 of 1.5"x 1 x2.25" (38x25x57 mm) compartments arranged 10 across x 10 deep.