Learn More
Leica Biosystems TISSUE FILTRATION MEDIUM CS
Catalog No. NC2584057
Encompass_Preferred
Tissue Infiltration Medium is a special formulation of purified paraffin and plastic polymers for infiltration or embedding. Thorough impregnation of all types of tissue specimens can be achieved with either open or closed automatic processor systems. This medium can be used in conjunction with EM-400.It is stable and will not break down or separate at low or high temperatures. Available in 2lb bags. Melting Point: 56-57° C. Water Bath Temperature: 45-50° C.
Catalog No. NC2584057 Supplier Leica Biosystems Supplier No. 3801340
Tissue Infiltration Medium is a special formulation of purified paraffin and plastic polymers for infiltration or embedding. Thorough impregnation of all types of tissue specimens can be achieved with either open or closed automatic processor systems. This medium can be used in conjunction with EM-400.It is stable and will not break down or separate at low or high temperatures. Available in 2lb bags. Melting Point: 56-57° C. Water Bath Temperature: 45-50° C.