Learn More
Cole-Parmer FB-200-115 Tissue Float Bath; 115 VAC, 50/60Hz
Catalog No. NC1191429 Shop All Cole-Parmer Products
Encompass_Preferred
Use the tissue float bath before the paraffin sections are placed on slides to prevent wrinkles and folds. The black PTFE interior and rim reduce eye strain by cutting down on glare while improving visibility of tissue sections. White powder-coated aluminum exterior is to keep clean. Safety features include an integrated indicator light alerting you when unit is on, as well as automatic shut-off.Order bath lid and a thermometer to help maintain desired temperature. Order gelatin to increase surface tension of water in flotation bath.Please Note: Effective May 2022, the Electrothermal tissue float bath Model number MH8517X1 has now been rebranded Cole-Parmer under Model number of FB-200-115. All the same great features and specifications, just with a new and improved look.
Catalog No. NC1191429 Supplier Cole-Parmer Supplier No. 4857250
Use the tissue float bath before the paraffin sections are placed on slides to prevent wrinkles and folds. The black PTFE interior and rim reduce eye strain by cutting down on glare while improving visibility of tissue sections. White powder-coated aluminum exterior is to keep clean. Safety features include an integrated indicator light alerting you when unit is on, as well as automatic shut-off.Order bath lid and a thermometer to help maintain desired temperature. Order gelatin to increase surface tension of water in flotation bath.Please Note: Effective May 2022, the Electrothermal tissue float bath Model number MH8517X1 has now been rebranded Cole-Parmer under Model number of FB-200-115. All the same great features and specifications, just with a new and improved look.