Learn More
World Precision Instrument Tissue Forceps, 14.5 cm, 3x4 Teeth
SDP
Catalog No. 502539499
Encompass
Tissue forceps with teeth have a fine tip with teeth for delicate tissue manipulation, such as during ophthalmic surgery or plastic surgery. Laboratory researchers choose these tissue forceps for precision work and microdissection. Choose between a 2x3, a 3x4, or a 4x5 tooth configuration. For example, 2x3 configuration has two teeth on one tip and three teeth on the other tip of the tweezers. The teeth ensure a firm grasp of the tissue, which is ideal for slippery or hard to hold tissues. While tweezers with teeth run the risk of puncturing delicate tissue, you do get a more secure hold. As a general rule, the fewer the teeth, the less likely the tissue trauma. These are available with straight tips. They are 14.5cm (5.7") long and fit easily into your hand for precision work.
Catalog No. 50-253-9499 Supplier World Precision Instrument Supplier No. 503312
Tissue forceps with teeth have a fine tip with teeth for delicate tissue manipulation, such as during ophthalmic surgery or plastic surgery. Laboratory researchers choose these tissue forceps for precision work and microdissection. Choose between a 2x3, a 3x4, or a 4x5 tooth configuration. For example, 2x3 configuration has two teeth on one tip and three teeth on the other tip of the tweezers. The teeth ensure a firm grasp of the tissue, which is ideal for slippery or hard to hold tissues. While tweezers with teeth run the risk of puncturing delicate tissue, you do get a more secure hold. As a general rule, the fewer the teeth, the less likely the tissue trauma. These are available with straight tips. They are 14.5cm (5.7") long and fit easily into your hand for precision work.