Learn More
Fine Science Tools Tissue Forceps, 1 x 2 Teeth, Stainless Steel, 15.5 cm
SDP
Catalog No. NC1653590
Encompass
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 2.5 x 1.7mm/ Alloy / Material: Stainless Steel/ Length: 15.5cm
This item is not returnable. View return policy
Catalog No. NC1653590 Supplier Fine Science Tools Supplier No. 1102115
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 2.5 x 1.7mm/ Alloy / Material: Stainless Steel/ Length: 15.5cm