Learn More
Med Vet International Miltex Vantage Tissue Froceps, 6in., With 1X2 Teeth
Catalog No. 502829497
Encompass_Preferred
The Miltex Vantage V96-46 Tissue Forceps are made of mid-grade stainless steel and measure 6in.They have 1x2 teeth that is used for grasping and securely holding tissue during surgical procedures while minimizing tissue damage.
Catalog No. 50-282-9497 Supplier Med Vet International Supplier No. MILV9646
The Miltex Vantage V96-46 Tissue Forceps are made of mid-grade stainless steel and measure 6in.They have 1x2 teeth that is used for grasping and securely holding tissue during surgical procedures while minimizing tissue damage.