Learn More
Electron Microscopy Sciences Tissue Forceps, Curved, PCF-FX
SDP
Catalog No. 5030640
Encompass_Preferred
12mm jaws with 1x2 teeth, very fine finish.
Model: PCF-FX
OAL: 140 mm
Catalog No. 50-306-40 Supplier Electron Microscopy Sciences Supplier No. 77898C
12mm jaws with 1x2 teeth, very fine finish.
Model: PCF-FX
OAL: 140 mm