Learn More
Med Vet International Miltex Vantage Tissue Forceps, 4.5in, with 1x2 Teeth
Supplier:  Med Vet International MILV9640
Encompass_Preferred
The Miltex Vantage V96-40 Tissue Forceps are made of mid-grade stainless steel and measure 4-5/8in.They have 1x2 teeth that is used for grasping and securely holding tissue during surgical procedures while minimizing tissue damage.
Catalog No. 50-255-9446