Learn More
Andwin Scientific TISSUE FREEZ. MEDIUM 12X4 OZ
SDP
Catalog No. 50949026
Encompass_Preferred
The Tissue Freezing Medium consists of ultra pure formulation of water-soluble glycols and resins. These components allow this product to offer a solid bond between the tissue and the object hold. It has a capacity of 4 x 4 ounces and has a yellow color.
Catalog No. 50-949-026 Supplier Andwin Scientific Supplier No. 72593
The Tissue Freezing Medium consists of ultra pure formulation of water-soluble glycols and resins. These components allow this product to offer a solid bond between the tissue and the object hold. It has a capacity of 4 x 4 ounces and has a yellow color.