Learn More
Leica Biosystems TISSUE FREEZING MED 125ML
Catalog No. NC0696746
Encompass
Tissue Freezing Medium is highly viscous and water soluble, making it an excellent choice for supporting tissue during cryotomy.Available in a 125 ml dripper bottle.
Catalog No. NC0696746 Supplier Leica Biosystems Supplier No. 14020108926
Tissue Freezing Medium is highly viscous and water soluble, making it an excellent choice for supporting tissue during cryotomy.Available in a 125 ml dripper bottle.