Learn More
Electron Microscopy Sciences TFM™ Tissue Freezing Medium, Blue 4 OZ
SDP
Catalog No. 50949017
Encompass_Preferred
An embedding matrix for frozen sections. TFM is an ultra pure formulation of water-soluble glycols and resins that provides a solid bond between the tissue and the object holder with the following features: less ice artifacts, less curling, and faster freezing.
Color: Blue
Catalog No. 50-949-017 Supplier Electron Microscopy Sciences Supplier No. 72592B
An embedding matrix for frozen sections. TFM is an ultra pure formulation of water-soluble glycols and resins that provides a solid bond between the tissue and the object holder with the following features: less ice artifacts, less curling, and faster freezing.
Color: Blue