Learn More
Electron Microscopy Sciences Tissue Grinder, 0.2 ml
SDP
Catalog No. 501900369
Encompass_Preferred
0.2 ml micro tissue grinder made from autoclavable Wheaton 33 low extractable borosilicate glass.
Size: 0.2 ml
Catalog No. 50-190-0369 Supplier Electron Microscopy Sciences Supplier No. 6479902
0.2 ml micro tissue grinder made from autoclavable Wheaton 33 low extractable borosilicate glass.
Size: 0.2 ml