Learn More
Chemglass Life Sciences Tissue Grinder, 10mL, Potter-Elvehjem, All Glass, 13mm Dia Pestle, 75mm Chember Lgth
SDP
Catalog No. 501532388
Encompass_Preferred
Tissue Grinder, 10mL, Potter-Elvehjem, All Glass, 13mm Dia Pestle, 75mm Chember Lgth. The rounded shape on the lower portion of the pestle progressively grinds the sample and the homogenate is produced as the sample is forced through the cylindrical section. Generally used with soft tissue such as brain and liver samples.
Catalog No. 50-153-2388 Supplier Chemglass Life Sciences Supplier No. CLS500405
Tissue Grinder, 10mL, Potter-Elvehjem, All Glass, 13mm Dia Pestle, 75mm Chember Lgth. The rounded shape on the lower portion of the pestle progressively grinds the sample and the homogenate is produced as the sample is forced through the cylindrical section. Generally used with soft tissue such as brain and liver samples.