Learn More
Chemglass Life Sciences Tissue Grinder, 10mL, Tenbroeck, 13mm Dia Pestle, 75mm Chamber Lgth, #00 Rubber Stopper
SDP
Catalog No. 501532399
Encompass_Preferred
Tissue Grinder, 10mL, Tenbroeck, 13mm Dia Pestle, 75mm Chamber Lgth, #00 Rubber Stopper. Are of the same design as the Potter Elvehjem style, but the pestle is hollow allowing the addition of ice water to keep samples chilled during the grinding process.
Catalog No. 50-153-2399 Supplier Chemglass Life Sciences Supplier No. CLS500602
Tissue Grinder, 10mL, Tenbroeck, 13mm Dia Pestle, 75mm Chamber Lgth, #00 Rubber Stopper. Are of the same design as the Potter Elvehjem style, but the pestle is hollow allowing the addition of ice water to keep samples chilled during the grinding process.