Learn More
Chemglass Life Sciences Tissue Grinder, 15mL, Style "A", Tapered, All Glass
SDP
Catalog No. 501532403
Encompass_Preferred
Tissue Grinder, 15mL, Style "A", Tapered, All Glass. The conical shape of this tissue grinder has a large surface for grinding making it the most efficient style of tissue grinder. Samples are progressively ground to a smaller size as the pestle is turned and lowered into the tube. It is used for tougher tissue such as heart, lung or muscle. Style A has a reservoir and a glass T handle.
Catalog No. 50-153-2403 Supplier Chemglass Life Sciences Supplier No. CLS500701
Tissue Grinder, 15mL, Style "A", Tapered, All Glass. The conical shape of this tissue grinder has a large surface for grinding making it the most efficient style of tissue grinder. Samples are progressively ground to a smaller size as the pestle is turned and lowered into the tube. It is used for tougher tissue such as heart, lung or muscle. Style A has a reservoir and a glass T handle.